Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193870.2 | NP_001180799.1 | 222 | 93 | ||
GenBank mRNA | JU319938.1 | - | 115 | 93 | ||
JU472920.1 | - | 199 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |