Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194448.2 | NP_001181377.1 | 1234 | 153 | ||
XM_015151627.2 | XP_015007113.1 | 1202 | 153 | |||
XM_028828933.1 | XP_028684766.1 | 2191 | 153 | |||
XM_028828934.1 | XP_028684767.1 | 1317 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |