Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090929.3 | XP_001090929.1 | 848 | 72 | ||
XM_028849955.1 | XP_028705788.1 | 818 | 72 | |||
XM_028849956.1 | XP_028705789.1 | 1339 | 72 | |||
GenBank mRNA | JV629713.1 | - | 840 | 72 | ||
JV697408.1 | - | 836 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |