Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR5V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194206.2 | NP_001181135.1 | 127 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |