Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106249.4 | XP_001106249.1 | 276 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |