Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092453.4 | XP_001092453.1 | 484 | 159 | ||
XM_015125773.2 | XP_014981259.1 | 304 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |