Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS2R40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080764.1 | NP_001074233.1 | 447 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |