Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261267.1 | NP_001248196.1 | 323 | 146 | ||
XM_015118222.2 | XP_014973708.2 | 275 | 146 | |||
XM_015118224.2 | XP_014973710.2 | 276 | 146 | |||
XM_028835545.1 | XP_028691378.1 | 276 | 146 | |||
GenBank mRNA | JU323017.1 | - | 323 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |