Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142265.2 | XP_014997751.1 | 5600 | 152 | ||
XM_015142266.2 | XP_014997752.1 | 5550 | 152 | |||
XM_015142267.2 | XP_014997753.1 | 5641 | 152 | |||
XM_015142268.2 | XP_014997754.1 | 5478 | 152 | |||
XM_015142269.2 | XP_014997755.1 | 5475 | 152 | |||
XM_015142270.2 | XP_014997756.1 | 5560 | 152 | |||
XM_015142271.2 | XP_014997757.1 | 5394 | 152 | |||
XM_028850935.1 | XP_028706768.1 | 5469 | 152 | |||
GenBank mRNA | AY369833.1 | - | 205 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |