Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091538.4 | XP_001091538.3 | 539 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |