Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001110286.4 | XP_001110286.2 | 1543 | 83 | ||
XM_015150160.2 | XP_015005646.2 | 1540 | 83 | |||
GenBank mRNA | JU323334.1 | - | 1448 | 83 | ||
JU476984.1 | - | 1336 | 83 | |||
JV452569.1 | - | 1336 | 83 | |||
JV567198.1 | - | 1470 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |