Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032814.1 | NP_001027986.1 | 279 | 160 | ||
GenBank mRNA | AF162280.1 | - | 279 | 160 | ||
JU322553.1 | - | 348 | 160 | |||
JV529872.1 | - | 346 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |