Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085477.4 | XP_001085477.2 | 413 | 138 | ||
XM_015135104.2 | XP_014990590.2 | 301 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |