Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150605.2 | XP_015006091.1 | 1065 | 75 | ||
GenBank mRNA | JU322603.1 | - | 663 | 75 | ||
JU471174.1 | - | 797 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |