Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104502.4 | XP_001104502.1 | 669 | 137 | ||
XM_015112036.2 | XP_014967522.1 | 830 | 137 | |||
XM_015112040.2 | XP_014967526.1 | 821 | 137 | |||
XM_028831805.1 | XP_028687638.1 | 2126 | 137 | |||
XM_028831806.1 | XP_028687639.1 | 1137 | 137 | |||
XM_028831807.1 | XP_028687640.1 | 1111 | 137 | |||
GenBank mRNA | JU326659.1 | - | 417 | 137 | ||
JV647302.1 | - | 605 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |