Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193925.3 | NP_001180854.2 | 1754 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |