Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PM20D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095121.4 | XP_001095121.1 | 900 | 101 | ||
GenBank mRNA | JU338943.1 | - | 290 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |