Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194394.2 | NP_001181323.1 | 745 | 132 | ||
XM_015126510.2 | XP_014981996.1 | 1106 | 132 | |||
XM_028840541.1 | XP_028696374.1 | 966 | 132 | |||
XM_028840543.1 | XP_028696376.1 | 1289 | 132 | |||
XM_028840544.1 | XP_028696377.1 | 830 | 132 | |||
GenBank mRNA | JV044015.1 | - | 781 | 132 | ||
JV635750.1 | - | 751 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |