Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001094500.4 | XP_001094500.2 | 423 | 66 | ||
GenBank mRNA | JU473596.1 | - | 391 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |