Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082856.4 | XP_001082856.1 | 1539 | 87 | ||
XM_015114701.2 | XP_014970187.1 | 1396 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |