Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC697943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001088476.4 | XP_001088476.1 | 1021 | 168 | ||
XM_015114076.2 | XP_014969562.1 | 1002 | 168 | |||
XM_015114077.2 | XP_014969563.1 | 831 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |