Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099694.3 | XP_001099694.2 | 2398 | 88 | ||
XM_001099902.3 | XP_001099902.2 | 2398 | 88 | |||
XM_015147968.2 | XP_015003454.2 | 2264 | 88 | |||
XM_015147986.2 | XP_015003472.2 | 2264 | 88 | |||
XM_015147996.2 | XP_015003482.2 | 2264 | 88 | |||
GenBank mRNA | JU327179.1 | - | 1041 | 88 | ||
JV636137.1 | - | 2145 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |