Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8H8orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144763.2 | XP_015000249.1 | 679 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |