Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC114675313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842554.1 | XP_028698387.1 | 627 | 142 |
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261809.1 | NP_001248738.1 | 874 | 142 | ||
XM_015145070.2 | XP_015000556.1 | 713 | 142 | |||
GenBank mRNA | JU319626.1 | - | 777 | 142 | ||
JU472900.1 | - | 817 | 142 | |||
JV044008.1 | - | 874 | 142 | |||
JV451657.1 | - | 892 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |