Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124271.2 | XP_014979757.1 | 1539 | 68 | ||
XM_015124272.2 | XP_014979758.1 | 1540 | 68 | |||
XM_015124273.2 | XP_014979759.1 | 1525 | 68 | |||
XM_015124275.2 | XP_014979761.1 | 1525 | 68 | |||
XM_015124276.2 | XP_014979762.1 | 1359 | 68 | |||
XM_015124277.2 | XP_014979763.1 | 1340 | 68 | |||
GenBank mRNA | AY369809.1 | - | 299 | 68 | ||
JV704488.1 | - | 553 | 68 | |||
JV704489.1 | - | 553 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |