Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266926.1 | NP_001253855.1 | 1023 | 96 | ||
XM_015126952.1 | XP_014982438.1 | 924 | 96 | |||
XM_015126953.2 | XP_014982439.1 | 1126 | 96 | |||
GenBank mRNA | JU320629.1 | - | 1023 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |