Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMU-DOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194694.2 | NP_001181623.1 | 693 | 125 | ||
GenBank mRNA | JU485413.1 | - | 652 | 125 | ||
JV704624.1 | - | 672 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta Inventoried |