Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258177.1 | NP_001245106.1 | 529 | 82 | ||
XM_015119212.2 | XP_014974698.1 | 438 | 82 | |||
GenBank mRNA | JU319721.1 | - | 529 | 82 | ||
JU319722.1 | - | 529 | 82 | |||
JU319723.1 | - | 529 | 82 | |||
JV046908.1 | - | 169 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |