Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261634.1 | NP_001248563.1 | 1500 | 66 | ||
XM_015146017.2 | XP_015001503.1 | 2065 | 66 | |||
XM_015146018.2 | XP_015001504.1 | 2208 | 66 | |||
XM_015146019.2 | XP_015001505.1 | 2631 | 66 | |||
XM_028852132.1 | XP_028707965.1 | 2488 | 66 | |||
XM_028852133.1 | XP_028707966.1 | 2484 | 66 | |||
XM_028852134.1 | XP_028707967.1 | 2356 | 66 | |||
XM_028852135.1 | XP_028707968.1 | 2409 | 66 | |||
XM_028852136.1 | XP_028707969.1 | 2368 | 66 | |||
GenBank mRNA | JU473118.1 | - | 1703 | 66 | ||
JU473119.1 | - | 1500 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |