Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194347.3 | NP_001181276.3 | 1534 | 146 | ||
XM_015112563.2 | XP_014968049.2 | 2111 | 146 | |||
XM_028831625.1 | XP_028687458.1 | 2042 | 146 | |||
GenBank mRNA | JU325969.1 | - | 452 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |