Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085459.4 | XP_001085459.3 | 1016 | 94 | ||
XM_015123477.2 | XP_014978963.2 | 579 | 94 | |||
XM_015123478.2 | XP_014978964.2 | 669 | 94 | |||
XM_015123480.2 | XP_014978966.2 | 548 | 94 | |||
XM_015123483.2 | XP_014978969.2 | 979 | 94 | |||
XM_028839928.1 | XP_028695761.1 | 852 | 94 | |||
GenBank mRNA | JV664417.1 | - | 688 | 94 | ||
JV699930.1 | - | 666 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |