Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266711.2 | NP_001253640.1 | 307 | 124 | ||
GenBank mRNA | JU321398.1 | - | 316 | 124 | ||
JU473147.1 | - | 323 | 124 | |||
JV046863.1 | - | 307 | 124 | |||
JV637252.1 | - | 326 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |