Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257440.1 | NP_001244369.1 | 723 | 140 | ||
GenBank mRNA | JU336692.1 | - | 723 | 140 | ||
JU477486.1 | - | 716 | 140 | |||
JV048434.1 | - | 719 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |