Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015121664.2 | XP_014977150.2 | 474 | 93 | ||
XM_015121665.2 | XP_014977151.2 | 474 | 93 | |||
XM_028837909.1 | XP_028693742.1 | 474 | 93 | |||
XM_028837912.1 | XP_028693745.1 | 345 | 93 | |||
XM_028837913.1 | XP_028693746.1 | 464 | 208 | |||
XM_028837914.1 | XP_028693747.1 | 349 | 208 | |||
GenBank mRNA | JV663985.1 | - | 452 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Macaca mulatta |