Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC52A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194561.2 | NP_001181490.1 | 1129 | 91 | ||
XM_015149297.2 | XP_015004783.1 | 1368 | 91 | |||
XM_015149298.2 | XP_015004784.1 | 1379 | 91 | |||
XM_015149299.2 | XP_015004785.1 | 1151 | 91 | |||
GenBank mRNA | JV046094.1 | - | 1308 | 91 | ||
JV635392.1 | - | 1371 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |