Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM207A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015132734.2 | XP_014988220.1 | 246 | 127 | ||
XM_015132735.2 | XP_014988221.1 | 246 | 127 | |||
GenBank mRNA | JU336829.1 | - | 139 | 127 | ||
JV048522.1 | - | 136 | 127 | |||
JV699484.1 | - | 128 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |