Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194162.2 | NP_001181091.1 | 451 | 115 | ||
GenBank mRNA | JU480179.1 | - | 107 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |