Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194428.3 | NP_001181357.2 | 515 | 112 | ||
XM_001108712.4 | XP_001108712.2 | 863 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |