Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194731.2 | NP_001181660.1 | 594 | 73 | ||
GenBank mRNA | JU332828.1 | - | 403 | 73 | ||
JU474239.1 | - | 522 | 73 | |||
JV046132.1 | - | 394 | 73 | |||
JV601388.1 | - | 441 | 73 | |||
JV638766.1 | - | 557 | 73 | |||
JV672360.1 | - | 412 | 73 |