Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HENMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257553.1 | NP_001244482.1 | 1017 | 130 | ||
XM_015146339.1 | XP_015001825.1 | 1057 | 130 | |||
XM_015146346.2 | XP_015001832.1 | 1030 | 130 | |||
XM_015146347.2 | XP_015001833.1 | 1051 | 130 | |||
XM_015146352.2 | XP_015001838.1 | 858 | 130 | |||
GenBank mRNA | JU332788.1 | - | 1017 | 130 | ||
JV046106.1 | - | 841 | 130 | |||
JV641442.1 | - | 905 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |