Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102240.4 | XP_001102240.1 | 1348 | 77 | ||
XM_001102336.4 | XP_001102336.1 | 1348 | 77 | |||
XM_028846475.1 | XP_028702308.1 | 1432 | 77 | |||
GenBank mRNA | JU330873.1 | - | 37 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |