Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193394.2 | NP_001180323.1 | 132 | 83 | ||
GenBank mRNA | JU323122.1 | - | 125 | 83 | ||
JU471916.1 | - | 132 | 83 | |||
JV045205.1 | - | 112 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |