Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114131.4 | XP_001114131.1 | 460 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |