Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCOCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119349.2 | XP_014974835.2 | 755 | 105 | ||
XM_028836399.1 | XP_028692232.1 | 648 | 105 | |||
GenBank mRNA | JU323336.1 | - | 712 | 105 | ||
JU473924.1 | - | 726 | 105 | |||
JV045360.1 | - | 696 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |