Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104263.4 | XP_001104263.1 | 1506 | 148 | ||
XM_015110295.2 | XP_014965781.1 | 1506 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |