Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266910.2 | NP_001253839.1 | 90 | 132 | ||
XM_015110132.2 | XP_014965618.1 | 292 | 132 | |||
XM_015110133.2 | XP_014965619.1 | 292 | 132 | |||
XM_015110134.2 | XP_014965620.1 | 292 | 132 | |||
GenBank mRNA | JU471922.1 | - | 90 | 132 | ||
JU471923.1 | - | 90 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |