Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190973.2 | NP_001177902.1 | 228 | 138 | ||
GenBank mRNA | JU320757.1 | - | 227 | 138 | ||
JU471088.1 | - | 233 | 138 | |||
JV044525.1 | - | 231 | 138 | |||
JV491241.1 | - | 147 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |