Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260665.1 | NP_001247594.1 | 942 | 93 | ||
XM_015149524.2 | XP_015005010.2 | 1046 | 93 | |||
XM_028827204.1 | XP_028683037.1 | 1008 | 93 | |||
GenBank mRNA | JU320037.1 | - | 966 | 93 | ||
JU471482.1 | - | 942 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |