Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093252.4 | XP_001093252.1 | 667 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |