Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092432.4 | XP_001092432.1 | 436 | 110 | ||
XM_002800090.3 | XP_002800136.1 | 544 | 110 | |||
XM_015117513.2 | XP_014972999.1 | 851 | 110 | |||
XM_015117514.2 | XP_014973000.1 | 848 | 110 | |||
XM_028835085.1 | XP_028690918.1 | 750 | 110 | |||
GenBank mRNA | JU333852.1 | - | 546 | 110 | ||
JV711454.1 | - | 350 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |